missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human pro-IGF2 Protein

Código de producto. p-7148658
Click to view available options
Cantidad:
10 μg
1 mg
2 μg
Tamaño de la unidad:
1 miligramo
10 microgramos
2 microgramos
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15905358

Marca: enQuireBio™ QP108482ug

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

A cDNA sequence encoding the pro-IGF2 was constructed and used to recombinantly synthesize the protein.

Specifications

Nombre pro-IGF2 Protein
Cantidad 2 μg
Estado normativo Research Use Only
Concentración de endotoxinas < 1.0 EU per ug protein as determined by the LAL method.
Actividad biológica The specific activity was determined by the dose-dependent stimulation of the proliferation of MCF-7 cells (human breast cancer cell line) and is typically 2-8ng/ml.
Tipo de producto Recombinant Protein
Reactividad cruzada Human
Especie Human (HEK293)
Etiqueta de proteína Untagged
Tampón The Pro-IGF2 was lyophilized in 50mM Sodium Acetate pH 4.5 and 350mM NaCl.
Grado de pureza o calidad Greater than 90% as obsereved by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.