Learn More
R&D Systems™ Recombinant Human Plexin D1 Protein
Descripción
Measured by its binding ability in a functional ELISA. When Recombinant Human Plexin-D1 is coated at 5μg/mL, Recombinant Human Semaphorin 3E (Catalog # 3239-S3 ) binds with an apparent Kd <10 nM.
The Recombinant Human Plexin D1 Protein is derived from NS0. The Recombinant Human Plexin D1 Protein has been validated for the following applications: Binding Activity.
Especificaciones
Especificaciones
| Formulación | Lyophilized from a 0.2μm filtered solution in PBS. |
| ID de gen (Entrez) | 23129 |
| Peso molecular | 132.7 kDa |
| Nombre | Plexin D1 |
| Cantidad | 50 μg |
| Fuente | Mouse myeloma cell line,NS0-derived human Plexin D1 protein Leu47-Ala1271 with a C-terminal 6-His tag |
| Requisitos de almacenamiento | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugado | Unconjugated |
| Recombinante | Recombinant |
| Grado de pureza o calidad | >85%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |