Learn More
enQuireBio™ Recombinant Human Oncostatin M / OSM Protein
Especificaciones
Especificaciones
| Número de acceso | NP_001306037.1 |
| ID de gen (Entrez) | 5008 |
| Nombre | Oncostatin M / OSM Protein |
| Cantidad | 500 μg |
| Estado normativo | Research Use Only |
| Concentración de endotoxinas | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
| Símbolo de gen | OSM |
| Actividad biológica | TF-1 cell proliferation was significantly induced by less than 4 ng/ml of Human Oncostatin M / OSM (minimum activity of at least 2.5 x 10^5 units/mg). |
| Tipo de producto | Recombinant Protein |
| Reactividad cruzada | Human |
| Mostrar más |