Learn More
enQuireBio™ Recombinant Human Non-specific lipid-transfer Protein
A DNA sequence encoding the Homo sapiens (Human) Non-specific lipid-transfer protein, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6653-ec-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001007099.1 | |
Non-specific lipid-transfer Protein | |
1 mg | |
Not determined. | |
Full Length of Isoform SCP2 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
6342 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
SCP2 | |
Recombinant Protein | |
E. coli | |
MGFPEAASSF RTHQIEAVPT SSASDGFKAN LVFKEIEKKL EEEGEQFVKK IGGIFAFKVK DGPGGKEATW VVDVKNGKGS VLPNSDKKAD CTITMADSDF LALMTGKMNP QSAFFQGKLK ITGNMGLAMK LQNLQLQPGN AKL |