Learn More
R&D Systems™ Recombinant Human IL-6 (HEK293-expressed) Protein
Código de producto. 18798926
Change view
Click to view available options
Cantidad:
100 μg
10 μg
Tamaño de la unidad:
10 microgramos
100 microgramos
18798926 100 μg 100 microgramos
18759036 10 μg 10 microgramos
2 options
Código de producto. 18798926
Marca: R&D Systems™ 7270IL100/CF
Measured in a cell proliferation assay using T1165.85.2.1 mouse plasmacytoma cells. ED50 for this effect is 0.1 to 0.6 ng/mL
Measured in a cell proliferation assay using T1165.85.2.1 mouse plasmacytoma cells. Nordan, R.P. et al. (1987) J. Immunol. 139:813. The ED50 for this effect is 0.1-0.6 ng/mL.
TRUSTED_SUSTAINABILITY
Especificaciones
Número de acceso P05231
Para utilizar con (aplicación) Bioactivity
Formulación Lyophilized from a 0.2 μm filtered solution in PBS.
ID de gen (Entrez) 3569
Peso molecular M.W. (Observed): 22 kDa, reducing conditions; M.W. (theoretical): 20.8 kDa
Nombre B cell stimulatory factor-2, B-cell differentiation factor, BSF2, BSF-2, BSF2CTL differentiation factor, CDF, HGFHSFIFNB2Hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6B-cell stimulatory factor 2, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2A
Cantidad 100 μg
Fuente Human embryonic kidney cell, HEK293-derived human IL-6 protein Met1-Met212
Requisitos de almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Concentración de endotoxinas <0.10 EU / 1 μg of the protein by the LAL method.
Alias de gen B-cell differentiation factor, B-cell stimulatory factor 2, BSF2, BSF-2, CDF, CTL differentiation factor, HSF, hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2A
Símbolo de gen IL6
Conjugado Unconjugated
Especie Human
Reconstitución Reconstitute at 100 μg/mL in PBS.
Tampón Lyophilized from a 0.2 μm filtered solution in PBS.
Nivel de endotoxinas <0.10 EU per 1 μg of the protein by the LAL method.
Grado de pureza o calidad >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Expresión IL-6