missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL 27 Protein

Product Code. 15994918
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Cantidad:
100 μg
1 μg
5 μg
missing translation for 'unitSize'
1 microgramo
100 microgramos
5 microgramos
Product Code. Cantidad unitSize
15994918 5 μg 5 microgramos
15984918 1 μg 1 microgramo
15974918 100 μg 100 microgramos
3 missing translation for 'options'
This item is not returnable. View return policy

Product Code. 15994918

missing translation for 'mfr': enQuireBio™ QP107065ug

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

A cDNA sequence encoding the IL 27 was constructed and used to recombinantly synthesize the protein.

Specifications

ID de gen (Entrez) 246778
Nombre IL 27 Protein
Cantidad 5 μg
Estado normativo Research Use Only
Concentración de endotoxinas < 1.0 EU per ug protein as determined by the LAL method.
Símbolo de gen IL27
Actividad biológica The specific activity was determined by the dose dependent inhibition of HIV replication in human CD4+ T cells and in human monocyte-derived macrophages and is typically 4-12ng/ml.
Tipo de producto Recombinant Protein
Reactividad cruzada Human
Especie Human (HEK293)
Etiqueta de proteína Untagged
Secuencia IL-27A (215aa):FPRPPGRPQL SLQELRREFT VSLHLARKLL AEVRGQAHRF AESHLPGVNL YLLPLGEQLP DVSLTFQAWR RLSDPERLCF ISTTLQPFHA LLGGLGTQGR WTNMERMQLW AMRLDLRDLQ RHLRFQVLAA GFNLPEEEEE EEEEEEEERK GLLPGALGSA LQGPAQVSWP QLLSTYRLLH SLELVLSRAV RELLLLSKAG HSVWPLGFPT LSPQP IL-27B (209aa):RKGPPAALTL PRVQCRASRY PIAVDCSWTL PPAPNSTSPV SFIATYRLGM AARGHSWPCL QQTPTSTSCT ITDVQLFSMA PYVLNVTAVH PWGSSSSFVP FITEHIIKPD PPEGVRLSPL AERQLQVQWE PPGSWPFPEI FSLKYWIRYK RQGAARFHRV GPIEATSFIL RAVRPRARYY VQVAAQDLTD YGELSDWSLP ATATMSLGK
Tampón The IL-27 was lyophilized from 1 mg/ml in 1xPBS.
Grado de pureza o calidad Greater than 90% as obsereved by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.