Learn More
R&D Systems™ Recombinant Human IL-23 (HEK293 Expressed) Protein
Código de producto. 18725755
Change view
Click to view available options
Cantidad:
100 μg
10 μg
1 mg
500 μg
50 μg
Tamaño de la unidad:
1 miligramo
10 microgramos
100 microgramos
50 microgramos
500 microgramos
18725755 100 μg 100 microgramos
18756185 500 μg 500 microgramos
18786065 50 μg 50 microgramos
18795875 1 mg 1 miligramo
18796215 10 μg 10 microgramos
5 options
Código de producto. 18725755
Marca: R&D Systems™ 11392IL100
Carrier Free Recombinant Protein
Measured by its ability to induce IL-17 secretion by mouse splenocytes. Aggarwal, S. et al. (2003) J. Biol. Chem. 278:1910. The ED50 for this effect is 0.100-2.00 ng/mL.
TRUSTED_SUSTAINABILITY
Especificaciones
Número de acceso P29460.1 & AAG37232.1
Formulación Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
ID de gen (Entrez) 51561
Peso molecular MolecularWeight-observed: 57-71 kDa, under reducing conditions., MolecularWeight-theroretical: 55 kDa
Cantidad 100 μg
Fuente Human embryonic kidney cell, HEK293-derived human IL-23 protein Human IL-12p40 (Ile23-Ser328) (N-terminus) Accession # P29460.1 GSGSSRGGSGSGGSGGGGSKL Human IL-23p19 (Arg20-Pro189) Accession # AAG37232.1 (C-terminus)
Requisitos de almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Concentración de endotoxinas <0.10 EU / 1 μg of the protein by the LAL method.
Alias de gen IL23, IL-23, IL-23 p19/IL-12 p40, IL-23 A, IL-23-A, IL-23p19, IL-23p19/IL-12p40, IL23P19P19, interleukin 23 p19 subunit, interleukin 23, alpha subunit p19, interleukin-23 subunit alpha, Interleukin-23 subunit p19, JKA3 induced upon T-cell activation, MGC79388, SGRF, SGRFIL-23 subunit alpha
Conjugado Unconjugated
Reconstitución Reconstitute 10 μg size at 100 μg/mL in PBS. Reconstitute all the other size (at 500 μg/mL in PBS.)
Grado de pureza o calidad >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Expresión IL-23 A/IL-23 P19
Mostrar más Mostrar menos
Name des Produkts
Wählen Sie ein Thema
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.