Learn More
DiscoverX Recombinant Human IL-11
Código de producto. 17322589
Encompass
Change view
Click to view available options
:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto.
17322589 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Código de producto. 17322589 Proveedor DiscoverX N.º de proveedor 921272
Recombinant Human IL-11 is an agonist for IL11RA. Provided as 10μg of dry powder alongside reconstitution buffer.
Interleukin 11 (IL-11) is a cytokine protein and mediator of the immune response. Stimulates T-cell dependent development of immunoglobulin-producing B cells. Stimulates proliferation of hematopoietic stem cells and megakaryocyte progenitor cells, inducing megakaryocyte maturation and platelet production. Agonist for IL11RA. Provided as 10μg of dry powder alongside reconstitution buffer.
Especificaciones
Cantidad 10 μg
Tipo de producto Control Ligands