missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human His6-Ubiquitin E1 Enzyme (UBE1), CF
Descripción
Recombinant Human His6-Ubiquitin Activating Enzyme (UBE1) is a member of the Ubiquitin-activating (E1) enzyme family that is required for the first step of the enzymatic cascade that subsequently utilizes a Ubiquitin-conjugating (E2) enzyme and a Ubiquitin ligase (E3) to conjugate Ubiquitin to substrate proteins. Reaction conditions will need to be optimized for each specific application. We recommend an initial Recombinant Human His6-Ubiquitin Activating Enzyme (UBE1) concentration of 50-200 nM.
Especificaciones
Especificaciones
| Número de acceso | P22314.3 |
| Para utilizar con (aplicación) | Enzyme Activity |
| Formulación | Supplied as a solution in HEPES, NaCl and TCEP. |
| ID de gen (Entrez) | 7317 |
| Peso molecular | MolecularWeight-theroretical: 121 kDa |
| Cantidad | 50 μg |
| Fuente | Spodoptera frugiperda, Sf 21 (baculovirus)-derived human Ubiquitin-activating Enzyme/UBE1 protein Contains an N-terminal Met-Ser-Tyr-Tyr and 6-His tag |
| Requisitos de almacenamiento | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
| Alias de gen | A1S9T, A1S9T and BN75 temperature sensitivity complementing, A1ST, AMCX1, GXP1, MGC4781, POC20, POC20 centriolar protein homolog, Protein A1S9, SMAX2, UBA1, UBA1, ubiquitin-activating enzyme E1 homolog A, UBA1A, UBE1, UBE1A1S9, UBE1X, Ubiquitinactivating Enzyme, Ubiquitin-activating enzyme E1, ubiquitin-activating enzyme E1 (A1S9T and BN75 temperature sensitivitycomplementing), ubiquitin-like modifier activating enzyme 1, ubiquitin-like modifier-activating enzyme 1 |
| Símbolo de gen | UBA1 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?