missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HINT1 Protein
Especificaciones
Especificaciones
| Nombre | HINT1 Protein |
| Cantidad | 5 μg |
| Estado normativo | Research Use Only |
| Concentración de endotoxinas | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Tipo de producto | Recombinant Protein |
| Reactividad cruzada | Human |
| Especie | E. coli |
| Etiqueta de proteína | Untagged |
| Secuencia | MADEIAKAQV ARPGGDTIFG KIIRKEIPAK IIFEDDRCLA FHDISPQAPT HFLVIPKKHI SQISVAEDDD ESLLGHLMIV GKKCAADLGL NKGYRMVVNE GSDGGQSVYH VHLHVLGGRQ MHWPPG |
| Tampón | The HINT1 solution contains 20mM Tris pH 8 and 10% glycerol. |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?