Learn More
enQuireBio™ Recombinant Human HDGF2 Protein
Código de producto. p-7150063
Change view
Click to view available options
Cantidad:
1 mg
20 μg
5 μg
Tamaño de la unidad:
1 miligramo
20 microgramos
5 microgramos
15999688 5 μg 5 microgramos
15989688 20 μg 20 microgramos
15979688 1 mg 1 miligramo
3 options
Código de producto. 15999688
Marca: enQuireBio™ QP122065ug
A cDNA sequence encoding the HDGF2 was constructed and used to recombinantly synthesize the protein.
Especificaciones
Nombre HDGF2 Protein
Cantidad 5 μg
Estado normativo Research Use Only
Concentración de endotoxinas Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Tipo de producto Recombinant Protein
Reactividad cruzada Human
Especie E. coli
Etiqueta de proteína His
Secuencia MGSSHHHHHH SSGLVPRGSH MGSHMARPRP REYKAGDLVF AKMKGYPHWP ARIDELPEGA VKPPANKYPI FFFGTHETAF LGPKDLFPYK EYKDKFGKSN KRKGFNEGLW EIENNPGVKF TGYQAIQQQS SSETEGEGGN TADASSEEEG DRVEEDGKGK RKNEKAGSKR KKSYTSKKSS KQSRKSPGDE DDKDCKEEEN KSSSEGGDAG NDTRNTTSDL QKTSEGT
Tampón HDGF2 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 10% glycerol and 0.1M NaCl.
Grado de pureza o calidad Greater than 90% as determined by SDS-PAGE.