missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GREM2 Protein
A cDNA sequence encoding the GREM2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12049-5ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
GREM2 Protein | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The GREM2 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |
Greater than 85% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSRKNRPAG AIPSPYKDGS SNNSERWQHQ IKEVLASSQE ALVVTERKYL KSDWCKTQPL RQTVSEEGCR SRTILNRFCY GQCNSFYIPR HVKKEEESFQ SCAFCKPQRV TSVLVELECP GLDPPFRLKK IQKVKQCRCM SVNLSDSDKQ |