Learn More
enQuireBio™ Recombinant Human GPD2 Protein
Código de producto. p-7149890
Change view
Click to view available options
Cantidad:
10 μg
1 mg
2 μg
Tamaño de la unidad:
1 miligramo
10 microgramos
2 microgramos
15999158 2 μg 2 microgramos
15979158 10 μg 10 microgramos
15989158 1 mg 1 miligramo
3 options
Código de producto. 15999158
Marca: enQuireBio™ QP120372ug
A cDNA sequence encoding the GPD2 was constructed and used to recombinantly synthesize the protein.
Especificaciones
ID de gen (Entrez) 2820
Nombre GPD2 Protein
Cantidad 2 μg
Estado normativo Research Use Only
Concentración de endotoxinas Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Símbolo de gen GPD2
Tipo de producto Recombinant Protein
Reactividad cruzada Human
Especie E. coli
Etiqueta de proteína His
Secuencia MAFQKAVKGT ILVGGGALAT VLGLSQFAHY RRKQMNLAYV KAADCISEPV NREPPSREAQ LLTLQNTSEF DILVIGGGAT GSGCALDAVT RGLKTALVER DDFSSGTSSR STKLIHGGVR YLQKAIMKLD IEQYRMVKEA LHERANLLEI APHLSAPLPI MLPVYKWWQL PYYWVGIKLY DLVAGSNCLK SSYVLSKSRA LEHFPMLQKD KLVGAIVYYD GQHNDARMNL AIALTAARYG AATANYMEVV SLLKKTDPQT GKVRVSGARC KDVLTGQEFD VRAKCVINAT GPFTDSVRKM DDKDAAAICQ PSAGVHIVMP GYYSPESMGL LDPATSDGRV IFFLPWQKMT IAGTTDTPTD VTHHPIPSEE DINFILNEVR NYLSCDVEVR RGDVLAAWSG IRPLVTDPKS ADTQSISRNH VVDISESGLI TIAGGKWTTY RSMAEDTINA AVKTHNLKAG PSRTVGLFLQ GGKDWSPTLY IRLVQDYGLE SEVAQHLAAT YGDKAFEVAK MASVTGKRWP IVGVRLVSEF PYIEAEVKYG IKEYACTAVD MISRRTRLAF LNVQAAEEAL PRIVELMGRE LNWDDYKKQE QLETARKFLY YEMGYKSRSE QLTDRSEISL LPSDIDRYKK RFHKFDADQK GFITIVDVQR VLESINVQMD ENTLHEILNE VDLNKNGQVE LNEFLQLMSA IQKGRVSGSR LAILMKTAEE NLDRRVPIPV DRSCGGL
Tampón GPD2 protein solution is supplied in 20mM Tris-HCl pH 8.0, 1mM EDTA and 50% glycerol.
Grado de pureza o calidad Greater than 95.0% as determined by SDS-PAGE.