missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GALK1 Protein
Especificaciones
Especificaciones
| Nombre | GALK1 Protein |
| Cantidad | 5 μg |
| Estado normativo | Research Use Only |
| Concentración de endotoxinas | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Tipo de producto | Recombinant Protein |
| Reactividad cruzada | Human |
| Especie | E. coli |
| Etiqueta de proteína | His |
| Secuencia | MGSSHHHHHH SSGLVPRGSH MAALRQPQVA ELLAEARRAF REEFGAEPEL AVSAPGRVNL IGEHTDYNQG LVLPMALELM TVLVGSPRKD GLVSLLTTSE GADEPQRLQF PLPTAQRSLE PGTPRWANYV KGVIQYYPAA PLPGFSAVVV SSVPLGGGLS SSASLEVATY TFLQQLCPDS GTIAARAQVC QQAEHSFAGM PCGIMDQFIS LMGQKGHALL IDCRSLETSL VPLSDPKLAV LITNSNVRHS LASSEYPVRR RQCEEVARAL GKESLREVQL EELEAARDLV SKEGFRRARH VVGEIRRTAQ AAAALRRGDY RAFGRLMVES HRSLRDDYEV SCPELDQLVE AALAVPGVYG SRMTGGGFGG CTVTLLEASA APHAMRHIQE HYGGTATFYL SQAADGAKVL CL |
| Tampón | GALK1 Human solution containing 20mM Tris pH 8.0, and 10% glycerol. |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?