Learn More
enQuireBio™ Recombinant Human FKBP5 Protein
Código de producto. p-7149735
Change view
Click to view available options
Cantidad:
10 μg
1 mg
2 μg
Tamaño de la unidad:
1 miligramo
10 microgramos
2 microgramos
15908698 2 μg 2 microgramos
15988688 10 μg 10 microgramos
15998688 1 mg 1 miligramo
3 options
Código de producto. 15908698
Marca: enQuireBio™ QP118862ug
A cDNA sequence encoding the FKBP5 was constructed and used to recombinantly synthesize the protein.
Especificaciones
ID de gen (Entrez) 2289
Nombre FKBP5 Protein
Cantidad 2 μg
Estado normativo Research Use Only
Concentración de endotoxinas Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Símbolo de gen FKBP5
Tipo de producto Recombinant Protein
Reactividad cruzada Human
Especie E. coli
Etiqueta de proteína His
Secuencia MKHHHHHHASMTTDEGAKNN EESPTATVAE QGEDITSKKD RGVLKIVKRV GNGEETPMIG DKVYVHYKGK LSNGKKFDSS HDRNEPFVFS LGKGQVIKAW DIGVATMKKG EICHLLCKPE YAYGSAGSLP KIPSNATLFF EIELLDFKGE DLFEDGGIIR RTKRKGEGYS NPNEGATVEI HLEGRCGGRM FDCRDVAFTV GEGEDHDIPI GIDKALEKMQ REEQCILYLG PRYGFGEAGK PKFGIEPNAE LIYEVTLKSF EKAKESWEMD TKEKLEQAAI VKEKGTVYFK GGKYMQAVIQ YGKIVSWLEM EYGLSEKESK ASESFLLAAF LNLAMCYLKL REYTKAVECC DKALGLDSAN EKGLYRRGEA QLLMNEFESA KGDFEKVLEV NPQNKAARLQ ISMCQKKAKE HNERDRRIYA NMFKKFAEQD AKEEANKAMG KKTSEGVTNE KGTDSQAMEE EKPEGHV
Tampón FKBP5 was filtered (0.4microm) and lyophilized in 20mM Tris buffer and 50mM NaCl, pH 7.5.
Grado de pureza o calidad Greater than 90.0% as determined by SDS-PAGE.