Learn More
enQuireBio™ Recombinant Human EDF1 Protein
Código de producto. p-7149584
Change view
Click to view available options
Cantidad:
1 mg
25 μg
5 μg
Tamaño de la unidad:
1 miligramo
25 microgramos
5 microgramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
15908228 5 μg 5 microgramos
15998218 25 μg 25 microgramos
15988218 1 mg 1 miligramo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 15908228 Proveedor enQuireBio™ N.º de proveedor QP117455ug
A cDNA sequence encoding the EDF1 was constructed and used to recombinantly synthesize the protein.
Especificaciones
Nombre EDF1 Protein
Cantidad 5 μg
Estado normativo Research Use Only
Concentración de endotoxinas Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Tipo de producto Recombinant Protein
Reactividad cruzada Human
Especie E. coli
Etiqueta de proteína His
Secuencia MAESDWDTVT VLRKKGPTAA QAKSKQAILA AQRRGEDVET SKKWAAGQNK QHSITKNTAK LDRETEELHH DRVTLEVGKV IQQGRQSKGL TQKDLATKIN EKPQVIADYE SGRAIPNNQV LGKIERAIGL KLRGKDIGKP IEKGPRAKLE HHHHHH
Tampón 0.5 mg/ml solution containing 20mM Tris pH 8.0, 1mM DTT, 0.1M NaCl and 20% glycerol.
Grado de pureza o calidad Greater than 90.0% as determined by SDS-PAGE.