missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Dystonin Protein
Especificaciones
Especificaciones
| Número de acceso | NP_001138241.1 |
| ID de gen (Entrez) | 667 |
| Nombre | Dystonin Protein |
| Cantidad | 500 μg |
| Estado normativo | Research Use Only |
| Concentración de endotoxinas | Not determined. |
| Símbolo de gen | DST |
| Longitud de la proteína | Partial (see sequence information for more details). |
| Tipo de producto | Recombinant Protein |
| Reactividad cruzada | Human |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?