missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human DUSP21 Protein

Código de producto. 15998118
Click to view available options
Cantidad:
10 μg
1 mg
2 μg
Tamaño de la unidad:
1 miligramo
10 microgramos
2 microgramos
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15998118

Marca: enQuireBio™ QP1171410ug

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

A cDNA sequence encoding the DUSP21 was constructed and used to recombinantly synthesize the protein.

Especificaciones

Nombre DUSP21 Protein
Cantidad 10 μg
Estado normativo Research Use Only
Concentración de endotoxinas Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Tipo de producto Recombinant Protein
Reactividad cruzada Human
Especie E. coli
Etiqueta de proteína His
Secuencia MGSSHHHHHH SSGLVPRGSH MGSHMTASAS SFSSSQGVQQ PSIYSFSQIT RSLFLSNGVA ANDKLLLSSN RITAIVNASV EVVNVFFEGI QYIKVPVTDA RDSRLYDFFD PIADLIHTID MRQGRTLLHC MAGVSRSASL CLAYLMKYHS MSLLDAHTWT KSRRPIIRPN NGFWEQLINY EFKLFNNNTV RMINSPVGNI PDIYEKDLRM MISM
Tampón The DUSP21 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 200mM NaCl, 2mM DTT and 20% glycerol.
Grado de pureza o calidad Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.