missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human DLX3 Protein

Código de producto. 15997988
Click to view available options
Cantidad:
1 mg
20 μg
5 μg
Unit Size:
1 miligramo
20 microgramos
5 microgramos
This item is not returnable. View return policy

Product Code. 15997988

Brand: enQuireBio™ QP116721mg

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

A cDNA sequence encoding the DLX3 was constructed and used to recombinantly synthesize the protein.

Specifications

Nombre DLX3 Protein
Cantidad 1 mg
Estado normativo Research Use Only
Concentración de endotoxinas Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Tipo de producto Recombinant Protein
Reactividad cruzada Human
Especie E. coli
Etiqueta de proteína His
Secuencia MGSSHHHHHH SSGLVPRGSH MGSMSGSFDR KLSSILTDIS SSLSCHAGSK DSPTLPESSV TDLGYYSAPQ HDYYSGQPYG QTVNPYTYHH QFNLNGLAGT GAYSPKSEYT YGASYRQYGA YREQPLPAQD PVSVKEEPEA EVRMVNGKPK KVRKPRTIYS SYQLAALQRR FQKAQYLALP ERAELAAQLG LTQTQVKIWF QNRRSKFKK
Tampón DLX3 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol.
Grado de pureza o calidad Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.