missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human d2hgdh, mitochondrial Protein

Código de producto. 16011591
Change view
Click to view available options
Cantidad:
100 μg
10 μg
1 mg
200 μg
500 μg
50 μg
Unit Size:
1 miligramo
10 microgramos
100 microgramos
200 microgramos
50 microgramos
500 microgramos
Product Code. Cantidad unitSize
16011591 1 mg 1 miligramo
16001591 10 μg 10 microgramos
16041591 50 μg 50 microgramos
16091581 100 μg 100 microgramos
16021591 200 μg 200 microgramos
16031591 500 μg 500 microgramos
6 options
This item is not returnable. View return policy

Product Code. 16011591

Brand: enQuireBio™ QP7919ec1mg

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) d2hgdh, mitochondrial, was expressed in the hosts and tags indicated.

Specifications

Número de acceso NP_001274178.1
ID de gen (Entrez) 728294
Nombre d2hgdh, mitochondrial Protein
Cantidad 1 mg
Estado normativo Research Use Only
Concentración de endotoxinas Not determined.
Símbolo de gen d2hgdh
Longitud de la proteína Full Length
Tipo de producto Recombinant Protein
Reactividad cruzada Human
Especie E. coli
Etiqueta de proteína His-SUMO
Secuencia LRGAPGAAGS WGRPVGPLAR RGCCSAPGTP EVPLTRERYP VRRLPFSTVS KQDLAAFERI VPGGVVTDPE ALQAPNVDWL RTLRGCSKVL LRPRTSEEVS HILRHCHERN LAVNPQGGNT GMVGGSVPVF DEIILSTARM NRVLSFHSVS GILVCQAGCV LEELSRYVEE RDFIMPLDLG AKGSCHIGGN VATNAGGLRF LRYGSLHGTV LGLEVVLADG TVLDCLTSLR KDNTGYDLKQ LFIGSEGTLG IITTVSILCP PKPRAVNVAF LGCPGFAEVL QTFSTCKGML GEILSAFEFM DAVCMQLVGR HLHLASPVQE SPFYVLIETS GSNAGHDAEK LGHFLEHALG SGLVTDGTMA TDQRKVKMLW ALRERITEAL SRDGYVYKYD LSLPVERLYD IVTDLRARLG PHAKHVVGYG HLGDGNLHLN VTAEAFSPSL LAALEPHVYE WTAGQQGSVS AEHGVGFRKR DVLGYSKPPG ALQLMQQLKA LLDPKGILNP YKTLPSQA
Tampón Tris-based buffer, 50% glycerol
Grado de pureza o calidad Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.