missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human CXCL11/I-TAC Protein
Descripción
Measured by its ability to chemoattract BaF3 mouse pro-B cells transfected with human CXCR3. The ED50 for this effect is 1-5 ng/mL.
Especificaciones
Especificaciones
| Formulación | Liofilizado a partir de una solución filtrada de 0,2 μm en PBS con BSA como proteína transportadora. |
| ID de gen (Entrez) | 6373 |
| Peso molecular | 8.3 kDa |
| Nombre | CXCL11/I-TAC |
| Cantidad | 25 μg |
| Fuente | E. coli-derived human CXCL11/I-TAC protein Phe22-Phe94 |
| Requisitos de almacenamiento | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugado | Unconjugated |
| Recombinante | Recombinant |
| Grado de pureza o calidad | >97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?