Learn More
enQuireBio™ Recombinant Human CD1A Protein
Código de producto. 15996788
Change view
Click to view available options
Cantidad:
1 μg
50 μg
5 μg
Tamaño de la unidad:
1 microgramo
5 microgramos
50 microgramos
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
15996788 5 μg 5 microgramos
15976788 1 μg 1 microgramo
15986788 50 μg 50 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Código de producto. 15996788 Proveedor enQuireBio™ N.º de proveedor QP113085ug
A cDNA sequence encoding the CD1A was constructed and used to recombinantly synthesize the protein.
Especificaciones
Nombre CD1A Protein
Cantidad 5 μg
Estado normativo Research Use Only
Concentración de endotoxinas Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Tipo de producto Recombinant Protein
Reactividad cruzada Human
Especie Baculovirus
Etiqueta de proteína His
Secuencia ADPDGLKEPL SFHVTWIASF YNHSWKQNLV SGWLSDLQTH TWDSNSSTIV FLCPWSRGNF SNEEWKELET LFRIRTIRSF EGIRRYAHEL QFEYPFEIQV TGGCELHSGK VSGSFLQLAY QGSDFVSFQN NSWLPYPVAG NMAKHFCKVL NQNQHENDIT HNLLSDTCPR FILGLLDAGK AHLQRQVKPE AWLSHGPSPG PGHLQLVCHV SGFYPKPVWV MWMRGEQEQQ GTQRGDILPS ADGTWYLRAT LEVAAGEAAD LSCRVKHSSL EGQDIVLYWE HHSSVHHHHH H
Tampón CD1A protein solution (0.5 mg/ml) containing Phosphate Buffered Saline (pH 7.4), and 10% glycerol.
Grado de pureza o calidad Greater than 85% as determined by SDS-PAGE.