missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CALML3 Protein

Product Code. 15987479
missing translation for 'orderingAttributeHoverText'
Cantidad:
100 μg
10 μg
1 mg
200 μg
500 μg
50 μg
missing translation for 'unitSize'
1 miligramo
10 microgramos
100 microgramos
200 microgramos
50 microgramos
500 microgramos
This item is not returnable. View return policy

Product Code. 15987479

missing translation for 'mfr': enQuireBio™ QP5748ec10ug

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) CALML3, was expressed in the hosts and tags indicated.

Specifications

Número de acceso NP_005176.1
ID de gen (Entrez) 810
Nombre CALML3 Protein
Cantidad 10 μg
Estado normativo Research Use Only
Concentración de endotoxinas Not determined.
Símbolo de gen CALML3
Longitud de la proteína Full Length
Tipo de producto Recombinant Protein
Reactividad cruzada Human
Especie E. coli
Etiqueta de proteína GST
Secuencia MADQLTEEQV TEFKEAFSLF DKDGDGCITT RELGTVMRSL GQNPTEAELR DMMSEIDRDG NGTVDFPEFL GMMARKMKDT DNEEEIREAF RVFDKDGNGF VSAAELRHVM TRLGEKLSDE EVDEMIRAAD TDGDGQVNYE EFVRVLVSK
Tampón Tris-based buffer, 50% glycerol
Grado de pureza o calidad Greater than 80% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.