missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human C1QBP Protein
A cDNA sequence encoding the C1QBP was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11229-5ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
C1QBP Protein | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The C1QBP protein solution contains 20mM Tris-HCl pH 7.5, 20% glycerol and 1mM DTT. |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MLHTDGDKAF VDFLSDEIKE ERKIQKHKTL PKMSGGWELE LNGTEAKLVR KVAGEKITVT FNINNSIPPT FDGEEEPSQG QKVEEQEPEL TSTPNFVVEV IKNDDGKKAL VLDCHYPEDE VGQEDEAESD IFSIREVSFQ STGESEWKDT NYTLNTDSLD WALYDHLMDF LADRGVDNTF ADELVELSTA LEHQEYITFL EDLKSFVKSQ |