missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human BUD31 Protein

Código de producto. p-7149034
Click to view available options
Cantidad:
100 μg
10 μg
2 μg
Tamaño de la unidad:
10 microgramos
100 microgramos
2 microgramos
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 15916508

Marca: enQuireBio™ QP112132ug

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

A cDNA sequence encoding the BUD31 was constructed and used to recombinantly synthesize the protein.

Especificaciones

ID de gen (Entrez) 8896
Nombre BUD31 Protein
Cantidad 2 μg
Estado normativo Research Use Only
Concentración de endotoxinas Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Símbolo de gen BUD31
Tipo de producto Recombinant Protein
Reactividad cruzada Human
Especie E. coli
Etiqueta de proteína His
Secuencia MGSSHHHHHH SSGLVPRGSH MGSMPKVKRS RKAPPDGWEL IEPTLDELDQ KMREAETEPH EGKRKVESLW PIFRIHHQKT RYIFDLFYKR KAISRELYEY CIKEGYADKN LIAKWKKQGY ENLCCLRCIQ TRDTNFGTNC ICRVPKSKLE VGRIIECTHC GCRGCSG
Tampón BUD31 protein solution (0.25 mg/ml) contains 20mM Tris-HCl buffer, (pH 8.0), 0.2M NaCl, 40% glycerol and 2mM DTT.
Grado de pureza o calidad Greater than 85.0% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.