Learn More
enQuireBio™ Recombinant Human BECN1 Protein
Código de producto. p-7148971
Click to view available options
Cantidad:
100 μg
10 μg
2 μg
Tamaño de la unidad:
10 microgramos
100 microgramos
2 microgramos
Código de producto. 15996308
Marca: enQuireBio™ QP111522ug
A cDNA sequence encoding the BECN1 was constructed and used to recombinantly synthesize the protein.
Especificaciones
ID de gen (Entrez) 8678
Nombre BECN1 Protein
Cantidad 2 μg
Estado normativo Research Use Only
Concentración de endotoxinas Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Símbolo de gen BECN1
Tipo de producto Recombinant Protein
Reactividad cruzada Human
Especie E. coli
Etiqueta de proteína His
Secuencia MGSSHHHHHH SSGLVPRGSH MEGSKTSNNS TMQVSFVCQR CSQPLKLDTS FKILDRVTIQ ELTAPLLTTA QAKPGETQEE ETNSGEEPFI ETPRQDGVSR RFIPPARMMS TESANSFTLI GEASDGGTME NLSRRLKVTG DLFDIMSGQT DVDHPLCEEC TDTLLDQLDT QLNVTENECQ NYKRCLEILE QMNEDDSEQL QMELKELALE EERLIQELED VEKNRKIVAE NLEKVQAEAE RLDQEEAQYQ REYSEFKRQQ LELDDELKSV ENQMRYAQTQ LDKLKKTNVF NATFHIWHSG QFGTINNFRL GRLPSVPVEW NEINAAWGQT VLLLHALANK MGLKFQRYRL VPYGNHSYLE SLTDKSKELP LYCSGGLRFF WDNKFDHAMV AFLDCVQQFK EEVEKGETRF CLPYRMDVEK GKIEDTGGSG GSYSIKTQFN SEEQWTKALK FMLTNLKWGL AWVSSQFYNK
Tampón BECN1 protein solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.5), 0.1M NaCl, 10% glycerol and 1mM DTT.
Grado de pureza o calidad Greater than 85.0% as determined by SDS-PAGE.