missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Bcl 6 Protein

Código de producto. 15996268
Change view
Click to view available options
Cantidad:
10 μg
2 μg
5 μg
missing translation for 'unitSize'
10 microgramos
2 microgramos
5 microgramos
Product Code. Cantidad unitSize
15996268 5 μg 5 microgramos
15986268 2 μg 2 microgramos
15976268 10 μg 10 microgramos
3 missing translation for 'options'
This item is not returnable. View return policy

Product Code. 15996268

missing translation for 'mfr': enQuireBio™ QP111395ug

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

A cDNA sequence encoding the Bcl 6 was constructed and used to recombinantly synthesize the protein.

Specifications

ID de gen (Entrez) 604
Nombre Bcl 6 Protein
Cantidad 5 μg
Estado normativo Research Use Only
Concentración de endotoxinas Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Símbolo de gen BCL6
Tipo de producto Recombinant Protein
Reactividad cruzada Human
Especie E. coli
Etiqueta de proteína GST
Secuencia MASPADSCIQ FTRHASDVLL NLNRLRSRDI LTDVVIVVSR EQFRAHKTVL MACSGLFYSI FTDQLKCNLS VINLDPEINP EGFCILLDFM YTSRLNLREG NIMAVMATAM YLQMEHVVDT CRKFIKASEA EMVSAIKPPR EEFLNSRMLM PQDIMAYRGR EVVENNLPLR SAPGCESRAF APSLYSGLST PPASYSMYSH LPVSSLLFSD EEFRDVRMPV ANPFPKERAL PCDSARPVPG EYSRPTLEVS PNVCHSNIYS PKETIPEEAR SDMHYSVAEG LKPAAPSARN APYFPCDKAS KEEERPSSED EIALHFEPPN APLNRKGLVS PQSPQKSDCQ PNSPTESCSS KNACILQASG SPPAKSPTDP KACNWKKYKF IVLNSLNQNA KPEGPEQAEL GRLSPRAYTA PPACQPPMEP ENLDLQSPTK LSASGEDSTI PQASRLNNIV NRSMTGSPRS SSESHSPLYM HPPKCTSCGS QSPQHAEMCL HTAGPTFPEE MGETQSEYSD SSCENGAFFC NECDCRFSEE ASLKRHTLQT HSDKPYKCDR CQASFRYKGN LASHKTVHTG EKPYRCNICG AQFNRPANLK THTRIHSGEK PYKCETCGAR FVQVAHLRAH VLIHTGEKPY PCEICGTRFR HLQTLKSHLR IHTGEKPYHC EKCNLHFRHK SQLRLHLRQK HGAITNTKVQ YRVSATDLPP ELPKAC
Tampón Bcl-6 protein in 50mM Tris-HCl, pH 7.5 and 10mM L-glutathione (reduced).
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.