Learn More
enQuireBio™ Recombinant Human APOC1 Protein
Código de producto. p-7148865
Click to view available options
Cantidad:
10 μg
1 mg
2 μg
Tamaño de la unidad:
1 miligramo
10 microgramos
2 microgramos
Código de producto. 15915988
Marca: enQuireBio™ QP110492ug
A cDNA sequence encoding the APOC1 was constructed and used to recombinantly synthesize the protein.
Especificaciones
ID de gen (Entrez) 341
Nombre APOC1 Protein
Cantidad 2 μg
Estado normativo Research Use Only
Concentración de endotoxinas Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Símbolo de gen APOC1
Tipo de producto Recombinant Protein
Reactividad cruzada Human
Especie E. coli
Etiqueta de proteína His
Secuencia MGSSHHHHHH SSGLVPRGSH MGSTPDVSSA LDKLKEFGNT LEDKARELIS RIKQSELSAK MREWFSETFQ KVKEKLKIDS
Tampón APOC1 protein solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl and 10% glycerol.
Grado de pureza o calidad Greater than 90% as determined by SDS-PAGE.