missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ANXA7 Protein
Specifications
Specifications
| Nombre | ANXA7 Protein |
| Cantidad | 20 μg |
| Estado normativo | Research Use Only |
| Concentración de endotoxinas | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Tipo de producto | Recombinant Protein |
| Reactividad cruzada | Human |
| Especie | E. coli |
| Etiqueta de proteína | His |
| Secuencia | MGSSHHHHHH SSGLVPRGSH MGSHMSYPGY PPTGYPPFPG YPPAGQESSF PPSGQYPYPS GFPPMGGGAY PQVPSSGYPG AGGYPAPGGY PAPGGYPGAP QPGGAPSYPG VPPGQGFGVP PGGAGFSGYP QPPSQSYGGG PAQVPLPGGF PGGQMPSQYP GGQPTYPSQP ATVTQVTQGT IRPAANFDAI RDAEILRKAM KGFGTDEQAI VDVVANRSND QRQKIKAAFK TSYGKDLIKD LKSELSGNME ELILALFMPP TYYDAWSLRK AMQGAGTQER VLIEILCTRT NQEIREIVRC YQSEFGRDLE KDIRSDTSGH FERLLVSMCQ GNRDENQSIN HQMAQEDAQR LYQAGEGRLG TDESCFNMIL ATRSFPQLRA TMEAYSRMAN RDLLSSVSRE FSGYVESGLK TILQCALNRP AFFAERLYYA MKGAGTDDST LVRIVVTRSE IDLVQIKQMF AQMYQKTLGT MIAGDTSGDY RRLLLAIVGQ |
| Tampón | ANXA7 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 150mM NaCl, 1mM DTT and 40% glycerol. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?