Learn More
R&D Systems™ Recombinant Human Annexin A6 Protein
Descripción
The Recombinant Human Annexin A6 Protein is derived from E. coli. The Recombinant Human Annexin A6 Protein has been validated for the following applications: Binding Activity.
Especificaciones
Especificaciones
| Formulación | Lyophilized from a 0.2μm filtered solution in PBS. |
| ID de gen (Entrez) | 309 |
| Peso molecular | 76.5 kDa |
| Nombre | Annexin A6 |
| Cantidad | 50μg |
| Fuente | E. coli-derived human Annexin A6 protein Ala2-Asp673,with a C-terminal 6-His tag |
| Requisitos de almacenamiento | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugado | Unconjugated |
| Recombinante | Recombinant |
| Grado de pureza o calidad | 85%, by SDS-PAGE under reducing conditions and visualized by silver stain. |