missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Amelogenin, X isoform Protein
Specifications
Specifications
| Número de acceso | NP_001133.1 |
| ID de gen (Entrez) | 265 |
| Nombre | Amelogenin, X isoform Protein |
| Cantidad | 50 μg |
| Estado normativo | Research Use Only |
| Concentración de endotoxinas | Not determined. |
| Símbolo de gen | AMELX |
| Longitud de la proteína | Full Length |
| Tipo de producto | Recombinant Protein |
| Reactividad cruzada | Human |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?