Learn More
enQuireBio™ Recombinant Human Allograft inflammatory factor 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Allograft inflammatory factor 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5630-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001305899.1 | |
Allograft inflammatory factor 1 Protein | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
199 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
Aif1 | |
Recombinant Protein | |
E. coli | |
SQTRDLQGGK AFGLLKAQQE ERLDEINKQF LDDPKYSSDE DLPSKLEGFK EKYMEFDLNG NGDIDIMSLK RMLEKLGVPK THLELKKLIG EVSSGSGETF SYPDFLRMML GKRSAILKMI LMYEEKAREK EKPTGPPAKK AISELP |