Learn More
enQuireBio™ Recombinant Human ACAT1 Protein
Código de producto. 15905628
Change view
Click to view available options
Cantidad:
1 mg
20 μg
5 μg
Tamaño de la unidad:
1 miligramo
20 microgramos
5 microgramos
15905628 5 μg 5 microgramos
15995618 20 μg 20 microgramos
15985618 1 mg 1 miligramo
3 options
Código de producto. 15905628
Marca: enQuireBio™ QP109365ug
A cDNA sequence encoding the ACAT1 was constructed and used to recombinantly synthesize the protein.
Especificaciones
Nombre ACAT1 Protein
Cantidad 5 μg
Estado normativo Research Use Only
Concentración de endotoxinas Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Tipo de producto Recombinant Protein
Reactividad cruzada Human
Especie E. coli
Etiqueta de proteína His
Secuencia MGSSHHHHHH SSGLVPRGSH MGSVSKPTLK EVVIVSATRT PIGSFLGSLS LLPATKLGSI AIQGAIEKAG IPKEEVKEAY MGNVLQGGEG QAPTRQAVLG AGLPISTPCT TINKVCASGM KAIMMASQSL MCGHQDVMVA GGMESMSNVP YVMNRGSTPY GGVKLEDLIV KDGLTDVYNKIHMGSCAENT AKKLNIARNE QDAYAINSYT RSKAAWEAGK FGNEVIPVTV TVKGQPDVVV KEDEEYKRVD FSKVPKLKTV FQKENGTVTA ANASTLNDGA AALVLMTADA AKRLNVTPLA RIVAFADAAV EPIDFPIAPV YAASMVLKDV GLKKEDIAMW EVNEAFSLVV LANIKMLEIDPQKVNINGGA VSLGHPIGMS GARIVGHLTH ALKQGEYGLA SICNGGGGAS AMLIQKL
Tampón The ACAT1 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 7.5), 0.1M NaCl, 10% glycerol and 1mM DTT.
Grado de pureza o calidad Greater than 95% as determined by SDS-PAGE.