Learn More
enQuireBio™ Recombinant Hepatitis C HCV NS5B Protein
A cDNA sequence encoding the HCV NS5B was constructed and used to recombinantly synthesize the protein.
248.00€ - 1815.00€
Especificaciones
Nombre Hepatitis C HCV NS5B Protein
Estado normativo Research Use Only
Concentración de endotoxinas Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Tipo de producto Recombinant Protein
Reactividad cruzada Hepatitis C
Ver más especificaciones
Productos 3
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
15999668
Ver documentos
enQuireBio™
QP12200-100ug
100 μg
248.00€
100 microgramos
15919678
Ver documentos
enQuireBio™
QP12200-500ug
500 μg
943.00€
500 microgramos
15909678
Ver documentos
enQuireBio™
QP12200-1mg
1 mg
1815.00€
1 miligramo
Especificaciones
Hepatitis C HCV NS5B Protein
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Hepatitis C
His
Protein is >95% pure as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
1.5M urea, 25mM Tris-HCl pH 8.0, 0.2% Triton-X and 50% Glycerol.
Documentos
Product Certifications
Título del producto
enQuireBio™ Recombinant Hepatitis C HCV NS5B Protein