missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Dog DMP1 Protein

Código de producto. 16016760
Change view
Click to view available options
Cantidad:
100 μg
10 μg
1 mg
200 μg
500 μg
50 μg
Tamaño de la unidad:
1 miligramo
10 microgramos
100 microgramos
200 microgramos
50 microgramos
500 microgramos
Código de producto. Cantidad unitSize
16016760 100 μg 100 microgramos
16026760 10 μg 10 microgramos
16066760 50 μg 50 microgramos
16046760 200 μg 200 microgramos
16056760 500 μg 500 microgramos
16036760 1 mg 1 miligramo
6 options
This item is not returnable. View return policy

Product Code. 16016760

Brand: enQuireBio™ QP6934ec100ug

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

A DNA sequence encoding the Canis lupus familiaris (Dog) (Canis familiaris) DMP1, was expressed in the hosts and tags indicated.

Specifications

Nombre DMP1 Protein
Cantidad 100 μg
Estado normativo Research Use Only
Concentración de endotoxinas Not determined.
Símbolo de gen DMP1
Longitud de la proteína Full Length
Tipo de producto Recombinant Protein
Reactividad cruzada Dog
Especie E. coli
Etiqueta de proteína His-SUMO
Secuencia MHRPAGGLSR SGGKEGDDKD DDEDDSGDDT FGDDDNGPGP EERQGGKSRL KSNEDSADST QSREDSPSLG EDSAQDTTSE SRDLDNEDEM NSSPERGDSV QESESKEHWV GGGSEGDSSH GDGSEFGDEE MQNDDPDTIR NERGNSRMNS ASVKSEESKG NSDEQASTQD SGDSQSMEYP RRKIFRKSRI SEEDDIGELD DSNTMEEVKS DSTENANSKE TGLSQSRENS ESESVEESEE NQSQEDSQNI QDPSSESSQE VDLPSQENSS ESQEEVVTES RGDNPDNTTS HSDQEDSDSS EENSLNKPSS SESESTEEQA DSESNESLRS SEESPESTEE NSSSQEDLQS HSASAESQSE ESQSEQDSQS EEDDASDSQD SSKLKEDSNS TESKSSSEEE GQSKNTEIES RKLIVDTYHN KPIGDQDDND CQDGY
Tampón Tris-based buffer, 50% glycerol
Grado de pureza o calidad Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.