missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dengue Virus Dengue Envelope-1 & 4 Protein
Specifications
Specifications
| Nombre | Dengue Envelope-1 & 4 Protein |
| Cantidad | 500 μg |
| Estado normativo | Research Use Only |
| Concentración de endotoxinas | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Tipo de producto | Recombinant Protein |
| Reactividad cruzada | Dengue Virus |
| Especie | E. coli |
| Etiqueta de proteína | His |
| Tampón | Phosphate buffered saline, pH 7.4. |
| Grado de pureza o calidad | Protein is >95% pure as determined by 12% PAGE (coomassie staining). |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?