missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Cyno Lymphotoxin beta R/TNFRSF3 His Protein
Descripción
Measured by its ability to inhibit Recombinant Human Lymphotoxin alpha 1/beta 2 (Catalog # 8884-LY ) induced IL-8 secretion in A375 human melanoma cells. The ED50 for this effect is 0.125-1.50 μg/mL.
Especificaciones
Especificaciones
| Número de acceso | XP_005569951.1 |
| Para utilizar con (aplicación) | Bioactivity |
| Formulación | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| ID de gen (Entrez) | 4055 |
| Peso molecular | M.W.-observed: 29-41 kDa, under reducing conditions., M.W.-theroretical: 26 kDa |
| Cantidad | 100 μg |
| Fuente | Chinese Hamster Ovary cell line, CHO-derived cynomolgus monkey Lymphotoxin beta R/TNFRSF3 protein Ser28-Met227, with a C-terminal 6-His tag |
| Requisitos de almacenamiento | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Concentración de endotoxinas | <0.10 EU / 1 μg of the protein by the LAL method. |
| Alias de gen | CD18, D12S370, ltbetar, LT-BETA-R, LTBR, lymphotoxin B receptor, lymphotoxin beta receptor (TNFR superfamily, member 3), Lymphotoxin-beta receptor, LymphotoxinbR, TNF RIII, TNF Rrp, TNFCRTNF-RIII, TNFR superfamily, member 3, TNFR2-RP, TNFR3, TNF-R-III, TNFR-RP, TNFRSF3, TNFRSF3TNFR-III, Tumor necrosis factor C receptor, Tumor necrosis factor receptor 2-related protein, tumor necrosis factor receptor superfamily member 3, tumor necrosis factor receptor superfamily, member 3, Tumor necrosis factor receptor type III |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?