missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Chlamydia HSP70 Protein
Specifications
Specifications
| Nombre | HSP70 Protein |
| Cantidad | 100 μg |
| Estado normativo | Research Use Only |
| Concentración de endotoxinas | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Tipo de producto | Recombinant Protein |
| Reactividad cruzada | Chlamydia |
| Especie | E. coli |
| Etiqueta de proteína | Untagged |
| Tampón | 50mM Tris-HCl, pH 8.0, 8M Urea, 60 mM NaCl and 50% glycerol. |
| Grado de pureza o calidad | Chlamydia HSP70 protein is >95% pure as determined by 10% PAGE (coomassie staining) and RP-HPLC. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?