missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine Vimentin Protein

Código de producto. 15995169
Click to view available options
Cantidad:
10 μg
1 mg
2 μg
missing translation for 'unitSize'
1 miligramo
10 microgramos
2 microgramos
This item is not returnable. View return policy

Product Code. 15995169

missing translation for 'mfr': enQuireBio™ QP139361mg

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

A cDNA sequence encoding the Vimentin was constructed and used to recombinantly synthesize the protein.

Specifications

Nombre Vimentin Protein
Cantidad 1 mg
Estado normativo Research Use Only
Concentración de endotoxinas Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Tipo de producto Recombinant Protein
Reactividad cruzada Bovine
Especie Bovine
Etiqueta de proteína Untagged
Secuencia MSTRSVSSSS YRRMFGGPGT ASRPSSTRSY VTTSTRTYSL GSALRPTTSR TLYTSSPGGV YATRSSAVRL RSGVPGVRLL QDSVDFSLAD AINTEFKNTR TNEKVELQEL NDRFANYIDK VRFLEQQNKI LLAELEQLKG QGKSRLGDLY EEEMRELRRQ VDQLTNDKAR VEVERDNLAE DIMRLREKLQ EEMLQREEAE STLQSFRQDV DNASLARLDL ERKVESLQEE IAFLKKLHDE EIQELQAQIQ EQHVQIDMDV SKPDLTAALR DVRQQYESVA AKNLQEAEEW YKSKFADLSE AANRNNDALR QAKQESNEYR RQVQTLTCEV DALKGTNESL ERQMREMEEN FSVEAANYQD TIGRLQDEIQ NMKEEMARHL REYQDLLNVK MALDIEIATY RKLLEGEESR ISLPLPNFSS LNLRETNLDS LPLVDTHSKR TLLIKTVETR DGQVINETSQ HHDDLE
Tampón The protein (1 mg/ml) was lyophilized from a 1 mg/ml solution containing 10mM sodium phosphate pH 7.5, 6M urea, 2mM DTT, 1mM EDTA and 10mM methylammonium chloride.
Grado de pureza o calidad Greater than 98.0% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.