missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Bovine Cytokeratin 17 Protein

Product Code. 16092130
Change view
Click to view available options
Cantidad:
100 μg
10 μg
1 mg
200 μg
500 μg
50 μg
Unit Size:
1 miligramo
10 microgramos
100 microgramos
200 microgramos
50 microgramos
500 microgramos
Product Code. Cantidad unitSize
16092130 100 μg 100 microgramos
16002140 10 μg 10 microgramos
16042140 50 μg 50 microgramos
16022140 200 μg 200 microgramos
16032140 500 μg 500 microgramos
16012140 1 mg 1 miligramo
6 options
This item is not returnable. View return policy

Product Code. 16092130

Brand: enQuireBio™ QP6277ec100ug

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

A DNA sequence encoding the Bos taurus (Bovine) Cytokeratin 17, was expressed in the hosts and tags indicated.

Specifications

Número de acceso NP_001098792.1
ID de gen (Entrez) 281889
Nombre Cytokeratin 17 Protein
Cantidad 100 μg
Estado normativo Research Use Only
Concentración de endotoxinas Not determined.
Símbolo de gen KRT17
Longitud de la proteína Full Length
Tipo de producto Recombinant Protein
Reactividad cruzada Bovine
Especie E. coli
Etiqueta de proteína His-SUMO
Secuencia MTTTIRHFSS GSIKGSSGLA GGSSRSCRVS GSLGGGSCRL GSAGGLGSGL GGSSYSSCYS FGSGGGYGSG GYVSGGYGGG FGGVDGLLVG GEKATMQNLN DRLASYLDKV RALEEANTEL ELKIRDWYQK QAPGPAPDYS SYFKTIEDLR NKIHTATVDN ANLLLQIDNA RLAADDFRTK FETEQALRVS VEADINGLRR VLDELTLARA DLEMQIENLK EELAYLRKNH EEEMKALRGQ VGGEINVEMD AAPGVDLSRI LNEMRDQYEK MAEKNRKDAE DWFFSKTEEL NREVATNSEL VQSGKSEISE LRRTLQALEI ELQSQLSMKA SLEGSLAETE NRYCMQLSQI QGLIGSVEEQ LAQLRCEMEQ QNQEYKILLD VKTRLEQEIA TYRRLLEGED AHLTQYKTKE PVTTRQVRTI VEEVQDGRVI SSREQVHQTS H
Tampón Tris-based buffer, 50% glycerol
Grado de pureza o calidad Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.