Learn More
RCAS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 12255-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This gene was identified as an estrogen-responsive gene. Regulation of transcription by estrogen is mediated by estrogen receptor which binds to the estrogen-responsive element found in the 5′-flanking region of this gene. The encoded protein is a tumor-associated antigen that is expressed at high frequency in a variety of cancers. Two transcript variants differing in the 5′ UTR, but encoding the same protein, have been identified for this gene.Especificaciones
| RCAS1 | |
| Polyclonal | |
| Unconjugated | |
| EBAG9 | |
| EB9, EBAG9, PDAF, RCAS1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 299864, 55960, 9166 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O00559, Q5PQP2, Q9D0V7 | |
| Ebag9 | |
| RCAS1 Fusion Protein Ag2905 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |