missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The RBM5 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Specifications
Specifications
| Línea de células | The lysate was created in HEK293T cells, using plasmid ID RC212543 and based on accession number NM_5778. The protein contains a C-terminal DDK tag. |
| Para utilizar con (aplicación) | Western Blot |
| Cantidad | 0,1 mg |
| Tampón de almacenamiento | RIPA buffer |
| Pureza | Protein |
| Especificidad de la prueba | Homo sapiens RNA binding motif protein 5 (RBM5), mRNA. |
| Contenido y almacenamiento | -80°C. Avoid freeze-thaw cycles. |
| Protein State | Native |
Para uso exclusivo en investigación
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?