Learn More
RBM4B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 15412-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
May play a role in alternative splice site selection during pre-mRNA processing.Especificaciones
| RBM4B | |
| Polyclonal | |
| Unconjugated | |
| RBM4B | |
| RBM30, RBM4B, RBM4L, RNA binding motif protein 30, RNA binding motif protein 4B, RNA binding protein 30, RNA binding protein 4B, ZCCHC15, ZCRB3B | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 474154, 66704, 83759 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.26 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q64LC9, Q8VE92, Q9BQ04 | |
| Rbm4b | |
| RBM4B Fusion Protein Ag7679 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |