missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
RBKS Polyclonal antibody specifically detects RBKS in Human samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Especificaciones
Especificaciones
| Antígeno | RBKS |
| Aplicaciones | ELISA, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Dilución | ELISA 1:5000-1:10000, Immunocytochemistry/ Immunofluorescence 1:300-1:10000, Immunohistochemistry-Paraffin 1:500-1:2000 |
| Formulación | 0.2 um filtered solution in PBS |
| Alias de gen | EC 2.7.1.15, RBSKDKFZp686G13268,5230400M11Rik, ribokinase |
| Especie del huésped | Rabbit |
| Inmunógeno | Produced in rabbits immunized with purified, recombinant Human RBKS (Accession#: NP_071411.1; Met1-Phe322) |
| Método de purificación | Antigen and protein A Affinity-purified |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?