Learn More
RBFOX3/NeuN Antibody (S09-7H2), Novus Biologicals™
Código de producto. 18675243 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μg
50 μg
Tamaño de la unidad:
100 microlitros
50 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
18675243 100 μg 100 microlitros
18675753 50 μg 50 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 18675243 Proveedor Novus Biologicals N.º de proveedor NBP319714100ul
Rabbit Monoclonal Antibody
RBFOX3/NeuN Monoclonal antibody specifically detects RBFOX3/NeuN in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Proximity Ligation Assay, Sandwich ELISA
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno RBFOX3/NeuN
Aplicaciones Western Blot, ELISA, Immunofluorescence, Proximity Ligation Assay, Sandwich ELISA
Clasificación Monoclonal
Clon S09-7H2
Concentración 0.3 mg/mL
Conjugado Unconjugated
Formulación 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Alias de gen FLJ56884, FLJ58356, Fox-1 homolog C, FOX3, FOX-3, hexaribonucleotide binding protein 3, HRNBP3, NeuN, neuN antigen, neuronal nuclei, neuronal nuclei antigen, RBFOX3, RNA binding protein fox-1 homolog 3, RNA binding protein, fox-1 homolog (C. elegans) 3, RNA binding protein, fox-1 homolog 3
Especie del huésped Rabbit
Inmunógeno A synthetic peptide of human RBFOX3/NeuN (Uniprot # A6NFN3)
Método de purificación Affinity purified
Cantidad 100 μg
Estado normativo RUO
Disciplina de investigación DNA replication Transcription Translation and Splicing, Neuronal Cell Markers, Neuroscience
Primario o secundario Primary
ID de gen (Entrez) 146713
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Formulario Purified
Isotype IgG