Learn More
Novus Biologicals™ Rat RBP4/Retinol-Binding Protein 4 ELISA Kit (Chemiluminescence)
Código de producto. 18620351 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
1 Kit
Tamaño de la unidad:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
18620351 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Código de producto. 18620351 Proveedor Novus Biologicals™ N.º de proveedor NBP268103
Kit (Chemiluminescence)
TRUSTED_SUSTAINABILITY
Especificaciones
Intervalo del ensayo De 0,31 a 20 ng/ml
Sensibilidad del ensayo 0.19 ng/mL
Tipo de producto ELISA Kit (Chemiluminescence)
Tipo de muestra Serum, Plasma, Biological Fluids
Especificidad This kit recognizes Rat RBP4 in samples. No significant cross-reactivity or interference between Rat RBP4 and analogues was observed.
Para utilizar con (aplicación) ELISA
ID de gen (Entrez) 5950
Símbolo de gen RBP4
Cantidad 1 Kit
Disciplina de investigación Cancer, Diabetes Research, Lipid and Metabolism
Requisitos de almacenamiento Storage of components varies. See protocol for specific instructions.
Sinónimo interstitial, Plasma retinol-binding protein, retinol binding protein 4, plasma, retinol-binding protein 4, plasma
Objetivo Rat