missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Rat M-CSF Recombinant Protein, PeproTech®
Descripción
Recombinant Rat M-CSF is a 36.2 kDa homodimeric protein consisting of two 155 amino acid polypeptide subunits. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
M-CSF (Macrophage colony-stimulating factor, CSF-1) is a survival factor essential for the proliferation and development of monocytes, macrophages, and osteoclast progenitor cells. M-CSF also induces VEGF (vascular endothelial growth factor) secretion by macrophages, thereby mediating mobilization of endothelial progenitor cells and neovascularization. M-CSF is present as several bioactive isoforms that differ in potency and stability. The full-length protein is synthesized as a membrane-spanning protein that can be expressed on the cell surface or further cleaved and modified in the secretory vesicle. Further, M-CSF is a disulfide-bonded homodimer which is processed into one of two isoforms, a glycoprotein or a proteoglycan that has been modified by the addition of chondroitin sulfate to each subunit. Binding of M-CSF to its receptor, c-Fms (CSF-1R or CD115) induces dimerization of the receptor followed by internalization and degradation of the complex. Functionally, M-CSF is known to stimulate differentiation of hematopoietic stem cells to monocyte-macrophage cell populations in culture. M-CSF acts through the CSF receptor 1. Although human M-CSF shows activity on mouse cells, mouse CSF shows no activity on human cells.
Especificaciones
Especificaciones
| Número de acceso | Q8JZQ0 |
| Para utilizar con (aplicación) | Functional Assay |
| Formulación | protein with no preservative |
| ID de gen (Entrez) | 78965 |
| Peso molecular | 36.2 kDa |
| Nombre | Rat M-CSF |
| Cantidad | 50 μg |
| Fuente | E. coli |
| Estado normativo | RUO |
| Concentración de endotoxinas | <1 EU/ μg |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?