Learn More
RASEF Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 28162-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Binds predomitly GDP, and also GTP.Especificaciones
| RASEF | |
| Polyclonal | |
| Unconjugated | |
| RASEF | |
| FLJ31614, RAB45, Ras related protein Rab 45, RASEF | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 158158, 298138 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8IZ41 | |
| RASEF | |
| RASEF Fusion Protein Ag28049 | |
| 150 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |