missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAP1A Antibody (5F8) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
203.00€ - 533.00€
Specifications
| Antígeno | RAP1A |
|---|---|
| Clon | 5F8 |
| Concentración | 1 mg/mL |
| Dilución | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready |
| Aplicaciones | Western Blot, Flow Cytometry, ELISA, CyTOF |
| Product Code | Brand | Cantidad | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Cantidad | Price | Quantity & Availability | |||||
|
18193589
|
Novus Biologicals
NBP2-37496 |
0.1 mL |
533.00€
0.10 ml |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
|
18689205
|
Novus Biologicals
NBP2-37496-0.025ml |
0.025 mL |
203.00€
0.03 ml |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
Description
RAP1A Monoclonal specifically detects RAP1A in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.Specifications
| RAP1A | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, CyTOF | |
| Unconjugated | |
| RUO | |
| Human | |
| C21KG, G-22K, GTP-binding protein smg p21A, KREV-1, KREV1Ras-related protein Krev-1, RAP1, RAP1A, member of RAS oncogene family, ras-related protein Rap-1A, RAS-related protein RAP1A, SMGP21 | |
| RAP1A | |
| IgG1 | |
| Affinity Purified |
| 5F8 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Cell Cycle and Replication, Endosome Markers | |
| PBS with 0.05% Sodium Azide | |
| 5906 | |
| Purified recombinant fragment of human RAP1A expressed in E. Coli. | |
| Primary | |
| 21 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title