missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Rad9 Monoclonal specifically detects Rad9 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Especificaciones
Especificaciones
| Antígeno | Rad9 |
| Aplicaciones | Western Blot, Immunohistochemistry |
| Clasificación | Monoclonal |
| Clon | OTI7B11 |
| Conjugado | Alexa Fluor 700 |
| Dilución | Western Blot, Immunohistochemistry |
| Formulación | 50mM Sodium Borate with 0.05% Sodium Azide |
| Alias de gen | cell cycle checkpoint control protein RAD9A, DNA repair exonuclease rad9 homolog A, EC 3.1.11.2, hRAD9, RAD9, RAD9 (S. pombe) homolog, RAD9 homolog A (S. pombe) |
| Símbolos de los genes | RAD9A |
| Especie del huésped | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?